missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-pyridinecarboxaldehyde, 97%, Thermo Scientific™

$98.89 - $233.43

Chemical Identifiers

CAS 34160-40-2
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.01
MDL Number MFCD02683546
InChI Key QWFHFNGMCPMOCD-UHFFFAOYSA-N
Synonym 6-bromo-2-pyridinecarboxaldehyde, 6-bromopicolinaldehyde, 6-bromo-2-formylpyridine, 6-bromopyridine-2-carboxaldehyde, 2-bromo-6-formylpyridine, 6-bromo-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-bromo, 2-bromopyridine-6-carboxaldehyde, 6-bromo-2-pyridine carboxaldehyde, 6-bromopicolylaldehyde
PubChem CID 2757009
IUPAC Name 6-bromopyridine-2-carbaldehyde
SMILES C1=CC(=NC(=C1)Br)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC387420010
View Documents Promotion Details
N/A 1g Glass bottle
Each for $98.89
Only null left
 
AC387420050
View Documents Promotion Details
N/A 5g Glass bottle
Each for $233.43
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

34160-40-2
186.01
QWFHFNGMCPMOCD-UHFFFAOYSA-N
2757009
C1=CC(=NC(=C1)Br)C=O
C6H4BrNO
MFCD02683546
6-bromo-2-pyridinecarboxaldehyde, 6-bromopicolinaldehyde, 6-bromo-2-formylpyridine, 6-bromopyridine-2-carboxaldehyde, 2-bromo-6-formylpyridine, 6-bromo-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-bromo, 2-bromopyridine-6-carboxaldehyde, 6-bromo-2-pyridine carboxaldehyde, 6-bromopicolylaldehyde
6-bromopyridine-2-carbaldehyde

Specifications

34160-40-2
100.0
96% min. (GC)
MFCD02683546
QWFHFNGMCPMOCD-UHFFFAOYSA-N
6-bromopyridine-2-carbaldehyde
2757009
97%
96.0
Authentic
C6H4BrNO
6-bromo-2-pyridinecarboxaldehyde, 6-bromopicolinaldehyde, 6-bromo-2-formylpyridine, 6-bromopyridine-2-carboxaldehyde, 2-bromo-6-formylpyridine, 6-bromo-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-bromo, 2-bromopyridine-6-carboxaldehyde, 6-bromo-2-pyridine carboxaldehyde, 6-bromopicolylaldehyde
C1=CC(=NC(=C1)Br)C=O
186.01
186.01
6-Bromo-2-pyridinecarboxaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only