missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-picoline, 97%

CAS: 5315-25-3 | C6H6BrN | 172.03 g/mol

$152.17 - $152.17

Chemical Identifiers

CAS 5315-25-3
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00040743
InChI Key SOHDPICLICFSOP-UHFFFAOYSA-N
Synonym 2-bromo-6-picoline, 6-bromo-2-picoline, 6-bromo-2-methylpyridine, 2-bromo-6-methyl-pyridine, 2-bromo-6-methyl pyridine, 2-brom-6-methylpyridin, pyridine, 2-bromo-6-methyl, 2-bromo-6-methy pyridine, 6-bromopicoline, 6-methyl-2-bromopyridine
PubChem CID 79205
IUPAC Name 2-bromo-6-methylpyridine
SMILES CC1=CC=CC(Br)=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC391050100
SDS View Documents
Thermo Scientific Chemicals
391050100
10 g Glass bottle
Each for $152.17
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5315-25-3
172.03
SOHDPICLICFSOP-UHFFFAOYSA-N
79205
CC1=CC=CC(Br)=N1
C6H6BrN
MFCD00040743
2-bromo-6-picoline, 6-bromo-2-picoline, 6-bromo-2-methylpyridine, 2-bromo-6-methyl-pyridine, 2-bromo-6-methyl pyridine, 2-brom-6-methylpyridin, pyridine, 2-bromo-6-methyl, 2-bromo-6-methy pyridine, 6-bromopicoline, 6-methyl-2-bromopyridine
2-bromo-6-methylpyridine

Specifications

5315-25-3
100.0
1.5120g/mL
97°C
96% min. (GC)
C6H6BrN
MFCD00040743
1.512
SOHDPICLICFSOP-UHFFFAOYSA-N
2-bromo-6-methylpyridine
79205
97%
6-Bromo-2-picoline
96.0
Colorless to Yellow
102°C to 103°C (20.0 mmHg)
Authentic
Glass bottle
1.5620 to 1.564
10 g
2-bromo-6-picoline, 6-bromo-2-picoline, 6-bromo-2-methylpyridine, 2-bromo-6-methyl-pyridine, 2-bromo-6-methyl pyridine, 2-brom-6-methylpyridin, pyridine, 2-bromo-6-methyl, 2-bromo-6-methy pyridine, 6-bromopicoline, 6-methyl-2-bromopyridine
CC1=CC=CC(Br)=N1
172.03
172.02
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-173-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only