missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-methylquinoline 98.0+%, TCI America™

Catalog No. p-7119908
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30395G 5 g
B303925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30395G Supplier TCI America Supplier No. B30395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 877-42-9
Molecular Formula C10H8BrN
Molecular Weight (g/mol) 222.085
MDL Number MFCD00079724
InChI Key SQRYQSKJZVQJAY-UHFFFAOYSA-N
Synonym 6-bromoquinaldine, 2-methyl-6-bromoquinoline, 6-bromo-2-methyl-quinoline, quinoline, 6-bromo-2-methyl, 6-bromo-2-methyl-1-azanaphthalene, zlchem 926, pubchem5882, acmc-209qps, ksc494e7l
PubChem CID 522885
IUPAC Name 6-bromo-2-methylquinoline
SMILES CC1=NC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 104°C
Color Yellow
Quantity 5 g
Formula Weight 222.09
Percent Purity ≥98.0% (GC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-methylquinoline