missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-methyl-3-nitropyridine, 97%

Catalog No. p-7055661
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6429906 5 g
AAH6429914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6429906 Supplier Thermo Scientific Chemicals Supplier No. H6429906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22282-96-8 | C6H5BrN2O2 | 217.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22282-96-8
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD03095219
InChI Key PCNVKBBKROTUNS-UHFFFAOYSA-N
Synonym 2-bromo-5-nitro-6-picoline, 2-bromo-6-methyl-5-nitropyridine, 2-bromo-5-nitro-6-methylpyridine, 6-bromo-2-methyl-3-nitro-pyridine, 6-bromo-3-nitro-2-picoline, 6-bromo-3-nitropicoline, 6-brom-2-methyl-3-nitro-pyridin, pubchem2968, acmc-209fuh, ksc549a9p
PubChem CID 24728014
IUPAC Name 6-bromo-2-methyl-3-nitropyridine
SMILES CC1=C(C=CC(Br)=N1)[N+]([O-])=O

Specifications

Melting Point 67°C to 72°C
Quantity 5 g
Beilstein 135841
Formula Weight 217.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromo-2-methyl-3-nitropyridine
Hazard Category H302-H315-H318-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only