missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-hydroxypyridine 98.0+%, TCI America™

Catalog No. p-7123521
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44081G 1 g
B44085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44081G Supplier TCI America Supplier No. B44081G

Chemical Identifiers

CAS 27992-32-1
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174.00
MDL Number MFCD00234043
InChI Key VOMMPWVMVDGZEM-UHFFFAOYSA-N
Synonym 6-Bromo-2-pyridinol, 6-Bromo-2(1H)-pyridinone, 6-Bromo-2-pyridone
PubChem CID 820497
IUPAC Name 6-bromo-1,2-dihydropyridin-2-one
SMILES BrC1=CC=CC(=O)N1

Specifications

Melting Point 123°C
Color Purple-White
Quantity 1 g
Formula Weight 174.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-hydroxypyridine