missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-chloroquinoline 98.0+%, TCI America™

Catalog No. p-7124146
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B56281G 1 g
B56285G 5 g
2 options

Catalog No. B56281G

Supplier: TCI America B56281G

Only null left

Chemical Identifiers

CAS 1810-71-5
Molecular Formula C9H5BrClN
Molecular Weight (g/mol) 242.50
MDL Number MFCD04115272
InChI Key YXRDWUJAJLDABJ-UHFFFAOYSA-N
PubChem CID 12894086
IUPAC Name 6-bromo-2-chloroquinoline
SMILES ClC1=NC2=CC=C(Br)C=C2C=C1

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 1 g
Synonym 6-bromo-2-chloro-quinoline, 2-chloro-6-bromoquinoline, quinoline, 6-bromo-2-chloro, pubchem14631, acmc-209ehe, 6-bromo-2 chloroquinoline, 2-chloro-6-bromo-quinoline, ksc537o9r, quinoline,6-bromo-2-chloro, abbypharma ap-14-10831
Formula Weight 242.50
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-chloroquinoline
TSCA No
Recommended Storage Refrigerator