missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-chloroquinoline, 96%

Catalog No. p-7036580
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5489503 1 g
AAH5489506 5 g
2 options

Catalog No. AAH5489503

Supplier: Thermo Scientific Chemicals H5489503

Only null left

CAS: 1810-71-5 | C9H5BrClN | 242.50 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in Chloroform and Methanol

Notes
Store at room temperature with proper ventilation.

Chemical Identifiers

CAS 1810-71-5
Molecular Formula C9H5BrClN
Molecular Weight (g/mol) 242.50
MDL Number MFCD04115272
InChI Key YXRDWUJAJLDABJ-UHFFFAOYSA-N
PubChem CID 12894086
IUPAC Name 6-bromo-2-chloroquinoline
SMILES ClC1=NC2=CC=C(Br)C=C2C=C1

Specifications

Melting Point 148°C to 150°C
Quantity 1 g
Synonym 6-bromo-2-chloro-quinoline, 2-chloro-6-bromoquinoline, quinoline, 6-bromo-2-chloro, pubchem14631, acmc-209ehe, 6-bromo-2 chloroquinoline, 2-chloro-6-bromo-quinoline, ksc537o9r, quinoline,6-bromo-2-chloro, abbypharma ap-14-10831
Solubility Information Soluble in Chloroform and Methanol
Formula Weight 242.5
Percent Purity 96%
Chemical Name or Material 6-Bromo-2-chloroquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only