missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2,4-dichloroquinazoline, 97%

Catalog No. p-7055108
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6360303 1 g
AAH6360306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6360303 Supplier Thermo Scientific Chemicals Supplier No. H6360303
Only null left

CAS: 102393-82-8 | C8H3BrCl2N2 | 277.93 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 102393-82-8
Molecular Formula C8H3BrCl2N2
Molecular Weight (g/mol) 277.93
MDL Number MFCD09744007
InChI Key LBAYOWRVZAKPLS-UHFFFAOYSA-N
PubChem CID 10107568
IUPAC Name 6-bromo-2,4-dichloroquinazoline
SMILES C1=CC2=C(C=C1Br)C(=NC(=N2)Cl)Cl

Specifications

Color White
Quantity 1 g
Synonym quinazoline, 6-bromo-2,4-dichloro, 2,4-dichloro-6-bromoquinazoline, 6-bromo-2,4-dichloro-quinazoline, quinazoline,6-bromo-2,4-dichloro, pubchem14690, acmc-1c6nj, abbypharma ap-16-10560, 6-bromanyl-2,4-bis chloranyl quinazoline
Solubility Information Slightly soluble in water.
Formula Weight 277.93
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromo-2,4-dichloroquinazoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only