missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2(1H)-quinolinone, 96%

Catalog No. p-7035874
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5409503 1 g
AAH5409506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5409503 Supplier Thermo Scientific Chemicals Supplier No. H5409503

CAS: 1810-66-8 | C9H6BrNO | 224.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1810-66-8
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.06
MDL Number MFCD08703169
InChI Key YLAFBGATSQRSTB-UHFFFAOYSA-N
Synonym 6-bromoquinolin-2 1h-one, 6-bromo-2 1h-quinolone, 6-bromo-2 1h-quinolinone, 6-bromoquinolin-2-ol, 6-bromo-2-quinolone, 6-bromo-2-hydroxyquinoline, 6-bromo-1,2-dihydroquinolin-2-one, 2 1h-quinolinone, 6-bromo, 2-hydroxy-6-bromoquinoline, 6-bromocarbostyril
PubChem CID 12378943
IUPAC Name 6-bromo-1H-quinolin-2-one
SMILES BrC1=CC=C2NC(=O)C=CC2=C1

Specifications

Melting Point >200°C
Quantity 1 g
Formula Weight 224.05
Percent Purity 96%
Chemical Name or Material 6-Bromo-2(1H)-quinolinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only