missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-indanone 98.0+%, TCI America™

Catalog No. B37115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37115G 5 g
B37111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37115G Supplier TCI America Supplier No. B37115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14548-39-1
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD02179286
InChI Key SEQHEDQNODAFIU-UHFFFAOYSA-N
Synonym 6-bromo-1-indanone, 6-bromoindanone, 6-bromo-2,3-dihydro-1h-inden-1-one, 6-bromo-indan-1-one, 6-bromoindan-1-one, 1-indanone, 6-bromo, 1h-inden-1-one, 6-bromo-2,3-dihydro, 6-bromo indanone, 6-bromo-indanone
PubChem CID 139778
IUPAC Name 6-bromo-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=CC(=C2)Br

Specifications

Melting Point 112°C
Quantity 5 g
Formula Weight 211.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-1-indanone