missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-hexanol 95.0+%, TCI America™

Catalog No. p-7119287
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B18225G 5 g
B182225G 25 g
2 options

Catalog No. B18225G

Supplier: TCI America B18225G

Only null left

Chemical Identifiers

CAS 4286-55-9
Molecular Formula C6H13BrO
Molecular Weight (g/mol) 181.07
MDL Number MFCD00002983
InChI Key FCMCSZXRVWDVAW-UHFFFAOYSA-N
Synonym 6-bromo-1-hexanol, 6-bromohexanol, 1-hexanol, 6-bromo, 6-bromo-hexan-1-ol, hexamethylene bromohydrin, 6-bromo-hexanol, 6-bromo-l-hexanol, 1-bromo-6-hexanol, 1-bromohexan-6-ol, 6-hydroxyhexyl bromide
PubChem CID 77970
IUPAC Name 6-bromohexan-1-ol
SMILES OCCCCCCBr

Specifications

Color Yellow
Boiling Point 114°C
Quantity 5 g
Formula Weight 181.07
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Bromo-1-hexanol
TSCA Yes
Recommended Storage Refrigerator