Learn More
6-Bromo-1-hexanol, 96%
CAS: 4286-55-9 | C6H13BrO | 181.07 g/mol
Supplier: Thermo Scientific Chemicals B2180306
Description
A product for proteomics research. Also used as a reagent in the preparation of nitric oxide-donating analogues of Sulindac (S699215), a non-steroidal anti-inflammatory drug that has the ability to prevent colon cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA product for proteomics research. Also used as a reagent in the preparation of nitric oxide-donating analogues of Sulindac (S699215), a non-steroidal anti-inflammatory drug that has the ability to prevent colon cancer.
Solubility
Soluble in chloroform and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 6-Bromo-1-hexanol | |
| 1.384 | |
| >110°C (230°F) | |
| 1.482 | |
| 5 g | |
| Soluble in chloroform and methanol. | |
| OCCCCCCBr | |
| 181.07 | |
| 181.08 |
| 4286-55-9 | |
| 105°C to 106°C (5 mmHg) | |
| C6H13BrO | |
| MFCD00002983 | |
| 6-bromo-1-hexanol, 6-bromohexanol, 1-hexanol, 6-bromo, 6-bromo-hexan-1-ol, hexamethylene bromohydrin, 6-bromo-hexanol, 6-bromo-l-hexanol, 1-bromo-6-hexanol, 1-bromohexan-6-ol, 6-hydroxyhexyl bromide | |
| FCMCSZXRVWDVAW-UHFFFAOYSA-N | |
| 6-bromohexan-1-ol | |
| 77970 | |
| 97% |
Chemical Identifiers
| 4286-55-9 | |
| 181.07 | |
| FCMCSZXRVWDVAW-UHFFFAOYSA-N | |
| 77970 | |
| OCCCCCCBr |
| C6H13BrO | |
| MFCD00002983 | |
| 6-bromo-1-hexanol, 6-bromohexanol, 1-hexanol, 6-bromo, 6-bromo-hexan-1-ol, hexamethylene bromohydrin, 6-bromo-hexanol, 6-bromo-l-hexanol, 1-bromo-6-hexanol, 1-bromohexan-6-ol, 6-hydroxyhexyl bromide | |
| 6-bromohexan-1-ol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only