missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-hexanol, 95%

Catalog No. AC270090025
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270090025 2.5 g Glass bottle
AC270090100 10 g Glass bottle
AC270090500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC270090025 Supplier Thermo Scientific Chemicals Supplier No. 270090025

CAS: 4286-55-9 | C6H13BrO | 181.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4286-55-9
Molecular Formula C6H13BrO
Molecular Weight (g/mol) 181.07
MDL Number MFCD00002983
InChI Key FCMCSZXRVWDVAW-UHFFFAOYSA-N
Synonym 6-bromo-1-hexanol, 6-bromohexanol, 1-hexanol, 6-bromo, 6-bromo-hexan-1-ol, hexamethylene bromohydrin, 6-bromo-hexanol, 6-bromo-l-hexanol, 1-bromo-6-hexanol, 1-bromohexan-6-ol, 6-hydroxyhexyl bromide
PubChem CID 77970
IUPAC Name 6-bromohexan-1-ol
SMILES OCCCCCCBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 1.3200g/mL
Boiling Point 104.0°C to 106.0°C (5.0 mmHg)
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4810 to 1.4840
Linear Formula Br(CH2)6OH
Quantity 2.5 g
Beilstein 01,1704
Specific Gravity 1.32
Formula Weight 181.07
Percent Purity 95%
Chemical Name or Material 6-Bromo-1-hexanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only