missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo[1,3]oxazolo[4,5-b]pyridin-2(3H)-one, 97%, Thermo Scientific Chemicals

Catalog No. p-4228462
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432680050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432680050 Supplier Thermo Scientific Chemicals Supplier No. 432680050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21594-52-5
Molecular Formula C6H3BrN2O2
Molecular Weight (g/mol) 215.01
MDL Number MFCD08689590
InChI Key VQPBRWIFFBIRRP-UHFFFAOYSA-N
Synonym 6-bromo-3h-oxazolo 4,5-b pyridin-2-one, 6-bromooxazolo 4,5-b pyridin-2 3h-one, 6-bromo-2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, oxazolo 4,5-b pyridin-2 3h-one, 6-bromo, 6-bromo 1,3 oxazolo 4,5-b pyridin-2 3h-one, 6-bromo-oxazolo 4,5-b pyridin-2 3h-one, 6-bromo-3h-1,3 oxazolo 4,5-b pyridin-2-one, pubchem17712, acmc-1cakb, 6-bromooxooxazolo 4,5-b pyridine
PubChem CID 13614458
IUPAC Name 6-bromo-3H-[1,3]oxazolo[4,5-b]pyridin-2-one
SMILES C1=C2C(=NC=C1Br)NC(=O)O2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 230°C to 232°C
Color Colorless to Amber
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 215.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Bromo[1, 3]oxazolo[4, 5-b]pyridin-2(3H)-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only