missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, Tech., Thermo Scientific™

Catalog No. CC27110EA
Change view
Click to view available options
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC27110EA Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC27110EA Supplier Thermo Scientific Chemicals Supplier No. CC27110EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 27452-17-1
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C14H19Br
Quantity 10g
MDL Number MFCD05664407
Synonym 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, naphthalene, 6-bromo-1,2,3,4-tetrahydro-1,1,4,4-tetramethyl, 1,1,4,4-tetramethyl-6-bromotetralin, 6-bromo-1,1,4,4-tetramethyl-tetralin, 6-bromo-1,1,4,4 tetramethy 1,2,3,4-tetrahydronaphthalene, 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphtalene, 2-bromo-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene, 2-bromo-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaph-thalene, 2-bromo-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphthalene
InChI Key NLOOVMVNNNYLFS-UHFFFAOYSA-N
SMILES CC1(CCC(C2=C1C=CC(=C2)Br)(C)C)C
IUPAC Name 6-bromo-1,1,4,4-tetramethyl-2,3-dihydronaphthalene
Molecular Weight (g/mol) 267.21
PubChem CID 226685
Formula Weight 267.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.