missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyloxyindole 98.0+%, TCI America™

Catalog No. B28091G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28091G 1 g
B28095G 5 g
B280925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B28091G Supplier TCI America Supplier No. B28091G

Chemical Identifiers

CAS 15903-94-3
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
InChI Key FPMICYBCFBLGOZ-UHFFFAOYSA-N
Synonym 6-benzyloxyindole, 6-benzyloxy-1h-indole, 1h-indole, 6-phenylmethoxy, 6-phenylmethoxy indole, 6-benzoxyindole, 6-benzyloxy-indole, 6-benzyloxy indole, 6-phenylmethoxyindole, pubchem7315
PubChem CID 260804
IUPAC Name 6-phenylmethoxy-1H-indole
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)C=CN3

Specifications

Melting Point 117°C
Quantity 1 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Benzyloxyindole