missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyloxyindole, 97%

Catalog No. AAL17805MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17805MD 250 mg
AAL1780503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17805MD Supplier Thermo Scientific Chemicals Supplier No. L17805MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15903-94-3 | C15H13NO | 223.275 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15903-94-3
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD00053554
InChI Key FPMICYBCFBLGOZ-UHFFFAOYSA-N
Synonym 6-benzyloxyindole, 6-benzyloxy-1h-indole, 1h-indole, 6-phenylmethoxy, 6-phenylmethoxy indole, 6-benzoxyindole, 6-benzyloxy-indole, 6-benzyloxy indole, 6-phenylmethoxyindole, pubchem7315
PubChem CID 260804
IUPAC Name 6-phenylmethoxy-1H-indole
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)C=CN3

Specifications

Melting Point 114°C to 116°C
Quantity 250 mg
Beilstein 173319
Formula Weight 223.28
Percent Purity 97%
Chemical Name or Material 6-Benzyloxyindole
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only