missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 135°C to 138°C |
| Quantity | 250 mg |
| Solubility Information | Insoluble in water. |
| Formula Weight | 310.2 |
| Percent Purity | 95% |
| Chemical Name or Material | 6-(Benzylamino)pyridine-3-boronic acid pinacol ester |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only