Learn More
6-Benzyladenine, 99%
CAS: 1214-39-7 | C12H11N5 | 225.26 g/mol
Supplier: Thermo Scientific Chemicals A1467806
Description
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsInhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.
Solubility
Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.
Notes
Incompatible with strong oxidizing agent.
Specifications
| 6-Benzyladenine | |
| 229°C to 233°C | |
| C12H11N5 | |
| 5 g | |
| 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap | |
| NWBJYWHLCVSVIJ-UHFFFAOYSA-N | |
| N-benzyl-7H-purin-6-amine | |
| 62389 | |
| 225.26 |
| 1214-39-7 | |
| 99% | |
| MFCD00005572 | |
| 19406 | |
| Soluble in water,methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane. | |
| C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1 | |
| 225.26 | |
| CHEBI:29022 | |
| 99% |
Chemical Identifiers
| 1214-39-7 | |
| 225.26 | |
| NWBJYWHLCVSVIJ-UHFFFAOYSA-N | |
| 62389 | |
| N-benzyl-7H-purin-6-amine |
| C12H11N5 | |
| MFCD00005572 | |
| 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap | |
| CHEBI:29022 | |
| C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-927-5
RTECSNumber : AU6252200
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only