missing translation for 'onlineSavingsMsg'
Learn More

6-Azathymine, 98%

Catalog No. p-7044494
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0676206 5 g
AAL0676214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0676206 Supplier Thermo Scientific Chemicals Supplier No. L0676206
Only null left

CAS: 932-53-6 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 932-53-6
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006457,MFCD00127802
InChI Key XZWMZFQOHTWGQE-UHFFFAOYSA-N
Synonym 6-azathymine, 6-methyl-1,2,4-triazine-3,5 2h,4h-dione, 5-methyl-6-azauracil, azathymine, usaf cb-28, 1,2,4-triazine-3,5 2h,4h-dione, 6-methyl, 6-methyl-1,2,4-triazine-3,5-diol, 6-methyl-1,2,4-triazine-3,5-dione, unii-zwx58t58za, azathymine, 6
PubChem CID 70269
SMILES CC1=NNC(=O)NC1=O

Specifications

Melting Point 211°C to 215°C
Quantity 5 g
Beilstein 126863
Merck Index 14,903
IUPAC Name 6-methyl-2H-1,2,4-triazine-3,5-dione
Formula Weight 127.1
Percent Purity 98%
Chemical Name or Material 6-Azathymine
EINECSNumber 213-253-9
RTECSNumber XY8050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only