missing translation for 'onlineSavingsMsg'
Learn More

6-Azaindole, 97+%

Catalog No. p-7055790
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6445803 1 g
AAH6445806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6445803 Supplier Thermo Scientific Chemicals Supplier No. H6445803

CAS: 271-29-4 | C7H6N2 | 118.14 g/mol

6-Azaindole is an indole derivative and useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Azaindole is an indole derivative and useful synthetic intermediate.

Solubility
Soluble in water (hardly), methanol and chloroform.

Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air.

Chemical Identifiers

CAS 271-29-4
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD01686899
InChI Key XLKDJOPOOHHZAN-UHFFFAOYSA-N
Synonym 6-azaindole, 1h-pyrrolo 2,3-c pyridine, harmyrine, 1,6-diazaindene, 6h-pyrrolo 2,3-c pyridine, 6-azaindol, pyrrolo 2,3-c pyridine, 6-aza-6h-indole, zlchem 439
PubChem CID 9219
IUPAC Name 1H-pyrrolo[2,3-c]pyridine
SMILES N1C=CC2=CC=NC=C12

Specifications

Quantity 1 g
Beilstein 109685
Sensitivity Air and light sensitive
Solubility Information Soluble in water (hardly),methanol and chloroform.
Formula Weight 118.14
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 6-Azaindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UY8720000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only