missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Aminoquinoxaline 98.0+%, TCI America™
$84.70 - $254.11
Chemical Identifiers
| CAS | 6298-37-9 |
|---|---|
| Molecular Formula | C8H7N3 |
| Molecular Weight (g/mol) | 145.17 |
| MDL Number | MFCD00462821 |
| InChI Key | MSGRFBKVMUKEGZ-UHFFFAOYSA-N |
| Synonym | 6-aminoquinoxaline, 6-quinoxalinamine, 6-quinoxalinylamine, quinoxalin-6-ylamine, quinoxaline-6-ylamine, 6-chinoxalinamin, 6-amino-quinoxaline, pubchem15453, quinoxalin, 6-amino |
| PubChem CID | 237859 |
| IUPAC Name | quinoxalin-6-amine |
| SMILES | NC1=CC=C2N=CC=NC2=C1 |
Chemical Identifiers
| 6298-37-9 | |
| 145.17 | |
| MSGRFBKVMUKEGZ-UHFFFAOYSA-N | |
| 237859 | |
| NC1=CC=C2N=CC=NC2=C1 |
| C8H7N3 | |
| MFCD00462821 | |
| 6-aminoquinoxaline, 6-quinoxalinamine, 6-quinoxalinylamine, quinoxalin-6-ylamine, quinoxaline-6-ylamine, 6-chinoxalinamin, 6-amino-quinoxaline, pubchem15453, quinoxalin, 6-amino | |
| quinoxalin-6-amine |
Specifications
| 6298-37-9 | |
| C8H7N3 | |
| 1 g | |
| MSGRFBKVMUKEGZ-UHFFFAOYSA-N | |
| quinoxalin-6-amine | |
| 237859 | |
| ≥98.0% (GC,T) | |
| 6-Aminoquinoxaline |
| 159°C | |
| MFCD00462821 | |
| 6-aminoquinoxaline, 6-quinoxalinamine, 6-quinoxalinylamine, quinoxalin-6-ylamine, quinoxaline-6-ylamine, 6-chinoxalinamin, 6-amino-quinoxaline, pubchem15453, quinoxalin, 6-amino | |
| NC1=CC=C2N=CC=NC2=C1 | |
| 145.17 | |
| 145.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No