missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoquinoxaline 98.0+%, TCI America™

Catalog No. A24901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24901G 1 g
A24905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24901G Supplier TCI America Supplier No. A24901G
Only null left

Chemical Identifiers

CAS 6298-37-9
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number MFCD00462821
InChI Key MSGRFBKVMUKEGZ-UHFFFAOYSA-N
Synonym 6-aminoquinoxaline, 6-quinoxalinamine, 6-quinoxalinylamine, quinoxalin-6-ylamine, quinoxaline-6-ylamine, 6-chinoxalinamin, 6-amino-quinoxaline, pubchem15453, quinoxalin, 6-amino
PubChem CID 237859
IUPAC Name quinoxalin-6-amine
SMILES NC1=CC=C2N=CC=NC2=C1

Specifications

Melting Point 159°C
Quantity 1 g
Formula Weight 145.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminoquinoxaline