missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoquinoline 99.0+%, TCI America™

Catalog No. A13701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13701G 1 g
A13705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13701G Supplier TCI America Supplier No. A13701G

Chemical Identifiers

CAS 580-15-4
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006803
InChI Key RJSRSRITMWVIQT-UHFFFAOYSA-N
Synonym 6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
PubChem CID 11373
IUPAC Name quinolin-6-amine
SMILES C1=CC2=C(C=CC(=C2)N)N=C1

Specifications

Melting Point 116°C
Boiling Point 187°C
Quantity 1 g
Formula Weight 144.18
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminoquinoline