missing translation for 'onlineSavingsMsg'
Learn More

6-Aminophthalide, 95%

Catalog No. p-7024364
Change view
Click to view available options
Quantity:
0.5 g
2.5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1889602 0.5 g
AAA1889688 2.5 g
AAA1889609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1889602 Supplier Thermo Scientific Chemicals Supplier No. A1889602
Only null left

CAS: 57319-65-0 | C8H7NO2 | 149.149 g/mol

It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.

Solubility
Soluble in water 1360 mg/L 25°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 57319-65-0
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00033530
InChI Key ZIJZDNKZJZUROE-UHFFFAOYSA-N
Synonym 6-aminophthalide, 6-aminoisobenzofuran-1 3h-one, 6-amino-1,3-dihydroisobenzofuran-1-one, 6-amino-2-benzofuran-1 3h-one, 6-amino-3h-isobenzofuran-1-one, 1 3h-isobenzofuranone, 6-amino, 6-amino-1,3-dihydro-2-benzofuran-1-one, 6-amino-1 3h-isobenzofuranone, 6-amino-3-hydroisobenzofuran-1-one, 6-aminophthalid
PubChem CID 93631
IUPAC Name 6-amino-3H-2-benzofuran-1-one
SMILES C1C2=C(C=C(C=C2)N)C(=O)O1

Specifications

Melting Point 182°C to 185°C
Quantity 0.5 g
Solubility Information Soluble in water 1360mg/L 25°C.
Formula Weight 149.15
Percent Purity 95%
Chemical Name or Material 6-Aminophthalide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 260-675-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only