missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinic Acid 98.0+%, TCI America™

Catalog No. p-7117468
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14861G 1 g
A14865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14861G Supplier TCI America Supplier No. A14861G
Only null left

Chemical Identifiers

CAS 3167-49-5
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00006326
InChI Key ZCIFWRHIEBXBOY-UHFFFAOYSA-N
Synonym 6-aminonicotinic acid, 3-pyridinecarboxylic acid, 6-amino, 2-amino-5-pyridinecarboxylic acid, 6-amino-nicotinic acid, 6-amino nicotinic acid, 2-aminopyridine-5-carboxylic acid, nicotinic acid, 6-amino, unii-9vr2o1ok60, 6-amino-3-pyridinecarboxylic acid, 6-aminopyridine 3-carboxylic acid
PubChem CID 18496
ChEBI CHEBI:68583
IUPAC Name 6-aminopyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)N

Specifications

Melting Point 312°C
Color White
Quantity 1 g
Formula Weight 138.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminonicotinic Acid