missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinic acid, 98%

CAS: 3167-49-5 | C6H6N2O2 | 138.13 g/mol

$227.91 - $227.91

Chemical Identifiers

CAS 3167-49-5
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00006326
InChI Key ZCIFWRHIEBXBOY-UHFFFAOYSA-N
Synonym 6-aminonicotinic acid, 3-pyridinecarboxylic acid, 6-amino, 2-amino-5-pyridinecarboxylic acid, 6-amino-nicotinic acid, 6-amino nicotinic acid, 2-aminopyridine-5-carboxylic acid, nicotinic acid, 6-amino, unii-9vr2o1ok60, 6-amino-3-pyridinecarboxylic acid, 6-aminopyridine 3-carboxylic acid
PubChem CID 18496
ChEBI CHEBI:68583
IUPAC Name 6-aminopyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC202540050
SDS View Documents
Thermo Scientific Chemicals
202540050
5 g Glass bottle
Each for $227.91
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3167-49-5
138.13
ZCIFWRHIEBXBOY-UHFFFAOYSA-N
18496
6-aminopyridine-3-carboxylic acid
C6H6N2O2
MFCD00006326
6-aminonicotinic acid, 3-pyridinecarboxylic acid, 6-amino, 2-amino-5-pyridinecarboxylic acid, 6-amino-nicotinic acid, 6-amino nicotinic acid, 2-aminopyridine-5-carboxylic acid, nicotinic acid, 6-amino, unii-9vr2o1ok60, 6-amino-3-pyridinecarboxylic acid, 6-aminopyridine 3-carboxylic acid
CHEBI:68583
C1=CC(=NC=C1C(=O)O)N

Specifications

3167-49-5
100.0
White to Beige
98%
C6H6N2O2
MFCD00006326
22, 542
6-aminonicotinic acid, 3-pyridinecarboxylic acid, 6-amino, 2-amino-5-pyridinecarboxylic acid, 6-amino-nicotinic acid, 6-amino nicotinic acid, 2-aminopyridine-5-carboxylic acid, nicotinic acid, 6-amino, unii-9vr2o1ok60, 6-amino-3-pyridinecarboxylic acid, 6-aminopyridine 3-carboxylic acid
ZCIFWRHIEBXBOY-UHFFFAOYSA-N
6-aminopyridine-3-carboxylic acid
18496
138.13
Powder
97.5
>300°C
Authentic
Glass bottle
H2N(C5H3N)COOH
5 g
15, 452
Solubility in water: ca 1.0g/L (20°C). Other solubilities: sparingly soluble in most solvents,5.0g/L (20°C) in ethanol
C1=CC(=NC=C1C(=O)O)N
138.13
CHEBI:68583
98%
6-Aminonicotinic acid, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 221-630-4

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only