missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinic acid, 98%

Catalog No. p-7081269
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1122306 5 g
AAA1122314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1122306 Supplier Thermo Scientific Chemicals Supplier No. A1122306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3167-49-5 | C6H6N2O2 | 138.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3167-49-5
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00006326
InChI Key ZCIFWRHIEBXBOY-UHFFFAOYSA-N
Synonym 6-aminonicotinic acid, 3-pyridinecarboxylic acid, 6-amino, 2-amino-5-pyridinecarboxylic acid, 6-amino-nicotinic acid, 6-amino nicotinic acid, 2-aminopyridine-5-carboxylic acid, nicotinic acid, 6-amino, unii-9vr2o1ok60, 6-amino-3-pyridinecarboxylic acid, 6-aminopyridine 3-carboxylic acid
PubChem CID 18496
ChEBI CHEBI:68583
IUPAC Name 6-aminopyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)N

Specifications

Melting Point ∼320°C (decomposition)
Quantity 5 g
Beilstein 115992
Merck Index 14,456
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 6-Aminonicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-630-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only