missing translation for 'onlineSavingsMsg'
Learn More

6-(Aminomethyl)quinoline 98.0+%, TCI America™

Catalog No. p-7118140
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2908200MG 200 mg
A29081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2908200MG Supplier TCI America Supplier No. A2908200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99071-54-2
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD02853688
InChI Key RZIPENSSTUBRAA-UHFFFAOYSA-N
Synonym 6-aminomethylquinoline, 6-quinolinemethanamine, 6-aminomethyl quinoline, quinolin-6-yl methanamine, quinolin-6-ylmethyl amine, quinoline-6-ylmethanamine, 1-quinolin-6-yl methanamine, 6-quinolylmethylamine, c-quinolin-6-yl-methylamine, 6-quinolylmethanamine
PubChem CID 1514384
IUPAC Name 1-(quinolin-6-yl)methanamine
SMILES NCC1=CC2=CC=CN=C2C=C1

Specifications

Boiling Point 159°C
Quantity 200 mg
Formula Weight 158.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 6-(Aminomethyl)quinoline