missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Aminomethyl)indole, 97%
Description
6-(Aminomethyl)indole is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-(Aminomethyl)indole is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
6-(Aminomethyl)indole is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 250 mg |
| Sensitivity | Moisture sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 146.19 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-(Aminomethyl)indole |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only