missing translation for 'onlineSavingsMsg'
Learn More

6-Aminohexanoic Acid 98.0+%, TCI America™

Catalog No. p-7116827
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A031225G 25 g
A0312500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A031225G Supplier TCI America Supplier No. A031225G

Chemical Identifiers

CAS 60-32-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00008238
InChI Key SLXKOJJOQWFEFD-UHFFFAOYSA-N
Synonym 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
PubChem CID 564
ChEBI CHEBI:16586
IUPAC Name 6-aminohexanoic acid
SMILES C(CCC(=O)O)CCN

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 25 g
Formula Weight 131.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminohexanoic Acid
EINECSNumber (9)-0069
RTECSNumber MO6300000
TSCA Yes