missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Aminohexanoic Acid 98.0+%, TCI America™
Supplier: TCI America A225525G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C6H13NO2 | |
| MFCD00008238 | |
| 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin | |
| CHEBI:16586 | |
| C(CCC(=O)O)CCN |
Specifications
| 6-Aminohexanoic Acid [for Biochemical Research] | |
| 204°C | |
| 25 g | |
| 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin | |
| C(CCC(=O)O)CCN | |
| 131.175 | |
| CHEBI:16586 | |
| ≥98.0% (T) |
| 60-32-2 | |
| C6H13NO2 | |
| MFCD00008238 | |
| SLXKOJJOQWFEFD-UHFFFAOYSA-N | |
| 6-aminohexanoic acid | |
| 564 | |
| 131.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0069
RTECSNumber : MO6300000
TSCA : Yes
Recommended Storage : Refrigerator