missing translation for 'onlineSavingsMsg'
Learn More

6-Aminocoumarin hydrochloride, 97%

Catalog No. AAL1116803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1116803 1 g
AAL1116806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1116803 Supplier Thermo Scientific Chemicals Supplier No. L1116803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 63989-79-7 | C9H8ClNO2 | 197.62 g/mol

6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.

Chemical Identifiers

CAS 63989-79-7
Molecular Formula C9H8ClNO2
Molecular Weight (g/mol) 197.62
MDL Number MFCD00082671
InChI Key OSIGAIXSSYAHEG-UHFFFAOYSA-N
Synonym 6-aminocoumarin hydrochloride, 6-amino-2h-chromen-2-one hydrochloride, 6-aminochromen-2-one hydrochloride, 2-oxochromen-6-yl azanium chloride, 6-aminocoumarin hci, acmc-20am07, 6-amino-chromen-2-one hydrochloride, 2-oxochromen-6-yl ammonium chloride, 6-amino-2h-chromen-2-one hydrochloride 1:1
PubChem CID 356789
IUPAC Name 6-aminochromen-2-one;hydrochloride
SMILES [Cl-].[NH3+]C1=CC=C2OC(=O)C=CC2=C1

Specifications

Melting Point >260°C (decomposition)
Quantity 1 g
Beilstein 3698955
Solubility Information Insoluble in water.
Formula Weight 197.62
Percent Purity 97%
Chemical Name or Material 6-Aminocoumarin hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber GN4930000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only