missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-5-bromoquinoline, 97%

Catalog No. AAH6315003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6315003 1 g
AAH6315006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6315003 Supplier Thermo Scientific Chemicals Supplier No. H6315003

CAS: 50358-42-4 | C9H7BrN2 | 223.073 g/mol

6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

Solubility
Slightly soluble in water. Soluble in alcohol, ether, and carbon disulfide.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 50358-42-4
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.073
MDL Number MFCD00234503
InChI Key MODLGTLYXJGDCH-UHFFFAOYSA-N
PubChem CID 12741232
IUPAC Name 5-bromoquinolin-6-amine
SMILES C1=CC2=C(C=CC(=C2Br)N)N=C1

Specifications

Color Yellow to Brown
Quantity 1 g
Synonym 6-amino-5-bromoquinoline, 6-quinolinamine, 5-bromo, 5-bromo-6-quinolinamine, 6-quinolinamine,5-bromo, 5-bromo-quinolin-6-ylamine, 6-quinolinamine, 5-bromo-9ci
Solubility Information Slightly soluble in water. Soluble in alcohol,ether,and carbon disulfide.
Formula Weight 223.07
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Amino-5-bromoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only