missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-3-chloro-1H-indazole, 97%
Description
6-Amino-3-chloro-1H-indazole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Amino-3-chloro-1H-indazole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Sparingly soluble in water (0.62 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
6-Amino-3-chloro-1H-indazole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Sparingly soluble in water (0.62 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 1 g |
| Solubility Information | Sparingly soluble in water (0.62g/L at 25°C). |
| Formula Weight | 167.6 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Amino-3-chloro-1H-indazole |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only