missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-methylquinoline 98.0+%, TCI America™

Catalog No. A10471G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10471G 1 g
A10475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10471G Supplier TCI America Supplier No. A10471G

Chemical Identifiers

CAS 65079-19-8
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00052600
InChI Key TYJFYUVDUUACKX-UHFFFAOYSA-N
Synonym 6-amino-2-methylquinoline, 2-methyl-6-quinolinamine, 6-quinolinamine, 2-methyl, 6-aminoquinaldine, 2-methyl-6-aminoquinoline, 6-quinolinamine,2-methyl, 2-methyl-quinolin-6-ylamine, 2-methyl-6-quinolylamine, mqn, 2-methylquinoline-6-amine
PubChem CID 103148
IUPAC Name 2-methylquinolin-6-amine
SMILES CC1=NC2=C(C=C1)C=C(C=C2)N

Specifications

Melting Point 189°C
Quantity 1 g
Formula Weight 158.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2-methylquinoline