Learn More
6-Amino-2-methylquinoline, 97%
CAS: 65079-19-8 | C10H10N2 | 158.204 g/mol
Supplier: Thermo Scientific Chemicals A13967MD
Description
6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 6-Amino-2-methylquinoline | |
| 189°C to 191°C | |
| MFCD00052600 | |
| 114795 | |
| Soluble in water. | |
| CC1=NC2=C(C=C1)C=C(C=C2)N | |
| 158.204 | |
| 158.2 |
| 65079-19-8 | |
| C10H10N2 | |
| 250 mg | |
| 6-amino-2-methylquinoline, 2-methyl-6-quinolinamine, 6-quinolinamine, 2-methyl, 6-aminoquinaldine, 2-methyl-6-aminoquinoline, 6-quinolinamine,2-methyl, 2-methyl-quinolin-6-ylamine, 2-methyl-6-quinolylamine, mqn, 2-methylquinoline-6-amine | |
| TYJFYUVDUUACKX-UHFFFAOYSA-N | |
| 2-methylquinolin-6-amine | |
| 103148 | |
| 97% |
Chemical Identifiers
| 65079-19-8 | |
| 158.204 | |
| TYJFYUVDUUACKX-UHFFFAOYSA-N | |
| 2-methylquinolin-6-amine |
| C10H10N2 | |
| MFCD00052600 | |
| 103148 | |
| CC1=NC2=C(C=C1)C=C(C=C2)N |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 000-000-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only