missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-2,4-lutidine 98.0+%, TCI America™
$88.56 - $286.85
Chemical Identifiers
| CAS | 5407-87-4 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.171 |
| MDL Number | MFCD00006322 |
| InChI Key | BRBUBVKGJRPRRD-UHFFFAOYSA-N |
| Synonym | 2-amino-4,6-dimethylpyridine, 6-amino-2,4-lutidine, 2-pyridinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyridinamine, 4,6-dimethyl-2-pyridylamine, pyridine, 2-amino-4,6-dimethyl, 2,4-lutidine, 6-amino, 2-amino-4,6-dimehtylpyridine, 4,6-dimethyl-2-aminopyridine, 2-amino-4,6-lutidine |
| PubChem CID | 21507 |
| IUPAC Name | 4,6-dimethylpyridin-2-amine |
| SMILES | CC1=CC(=NC(=C1)N)C |
Chemical Identifiers
| 5407-87-4 | |
| 122.171 | |
| BRBUBVKGJRPRRD-UHFFFAOYSA-N | |
| 21507 | |
| CC1=CC(=NC(=C1)N)C |
| C7H10N2 | |
| MFCD00006322 | |
| 2-amino-4,6-dimethylpyridine, 6-amino-2,4-lutidine, 2-pyridinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyridinamine, 4,6-dimethyl-2-pyridylamine, pyridine, 2-amino-4,6-dimethyl, 2,4-lutidine, 6-amino, 2-amino-4,6-dimehtylpyridine, 4,6-dimethyl-2-aminopyridine, 2-amino-4,6-lutidine | |
| 4,6-dimethylpyridin-2-amine |
Specifications
| 5407-87-4 | |
| White-Yellow | |
| C7H10N2 | |
| 5 g | |
| BRBUBVKGJRPRRD-UHFFFAOYSA-N | |
| 4,6-dimethylpyridin-2-amine | |
| 21507 | |
| ≥98.0% (GC,T) | |
| 6-Amino-2,4-lutidine |
| 67°C | |
| 231°C | |
| MFCD00006322 | |
| 2-amino-4,6-dimethylpyridine, 6-amino-2,4-lutidine, 2-pyridinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyridinamine, 4,6-dimethyl-2-pyridylamine, pyridine, 2-amino-4,6-dimethyl, 2,4-lutidine, 6-amino, 2-amino-4,6-dimehtylpyridine, 4,6-dimethyl-2-aminopyridine, 2-amino-4,6-lutidine | |
| CC1=CC(=NC(=C1)N)C | |
| 122.171 | |
| 122.17 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : US1818000
TSCA : Yes