missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-1-methyluracil 98.0+%, TCI America™
$160.30 - $160.30
Chemical Identifiers
| CAS | 2434-53-9 |
|---|---|
| Molecular Formula | C5H7N3O2 |
| Molecular Weight (g/mol) | 141.13 |
| MDL Number | MFCD00075366 |
| InChI Key | GZLZRPNUDBIQBM-UHFFFAOYSA-N |
| Synonym | 6-amino-1-methyluracil, 6-amino-1-methylpyrimidine-2,4 1h,3h-dione, 1-methyl-6-aminouracil, 2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl, 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-amino-1-methyl-2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl-1h-pyrimidine-2,4-dione, 6-amino-1-methyl-1,3-dihydropyrimidine-2,4-dione, 6-amino-1-methyl-3h-pyrimidine-2,4-dione, zlchem 437 |
| PubChem CID | 75520 |
| IUPAC Name | 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| SMILES | CN1C(=O)NC(=O)C=C1N |
Chemical Identifiers
| 2434-53-9 | |
| 141.13 | |
| GZLZRPNUDBIQBM-UHFFFAOYSA-N | |
| 75520 | |
| CN1C(=O)NC(=O)C=C1N |
| C5H7N3O2 | |
| MFCD00075366 | |
| 6-amino-1-methyluracil, 6-amino-1-methylpyrimidine-2,4 1h,3h-dione, 1-methyl-6-aminouracil, 2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl, 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-amino-1-methyl-2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl-1h-pyrimidine-2,4-dione, 6-amino-1-methyl-1,3-dihydropyrimidine-2,4-dione, 6-amino-1-methyl-3h-pyrimidine-2,4-dione, zlchem 437 | |
| 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione |
Specifications
| 2434-53-9 | |
| White | |
| MFCD00075366 | |
| 6-amino-1-methyluracil, 6-amino-1-methylpyrimidine-2,4 1h,3h-dione, 1-methyl-6-aminouracil, 2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl, 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-amino-1-methyl-2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl-1h-pyrimidine-2,4-dione, 6-amino-1-methyl-1,3-dihydropyrimidine-2,4-dione, 6-amino-1-methyl-3h-pyrimidine-2,4-dione, zlchem 437 | |
| CN1C(=O)NC(=O)C=C1N | |
| 141.13 | |
| 141.13 | |
| Crystalline Powder |
| 307°C | |
| C5H7N3O2 | |
| 5 g | |
| GZLZRPNUDBIQBM-UHFFFAOYSA-N | |
| 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 75520 | |
| ≥98.0% (HPLC,T) | |
| 6-Amino-1-methyluracil |
Safety and Handling
TSCA : No