missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-hexanol 97.0+%, TCI America™

Catalog No. p-7117198
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10275G 5 g
A102725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10275G Supplier TCI America Supplier No. A10275G
Only null left

Chemical Identifiers

CAS 4048-33-3
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00008241
InChI Key SUTWPJHCRAITLU-UHFFFAOYSA-N
Synonym 6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol
PubChem CID 19960
IUPAC Name 6-aminohexan-1-ol
SMILES C(CCCO)CCN

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 225°C
Quantity 5 g
Formula Weight 117.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1-hexanol
RTECSNumber MO8840000
TSCA No