missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-hexanol, 97%, Thermo Scientific Chemicals

Catalog No. p-7046389
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1415706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1415706 Supplier Thermo Scientific Chemicals Supplier No. L1415706

6-Amino-1-hexanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-1-hexanol is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 4048-33-3
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00008241
InChI Key SUTWPJHCRAITLU-UHFFFAOYSA-N
Synonym 6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol
PubChem CID 19960
IUPAC Name 6-aminohexan-1-ol
SMILES C(CCCO)CCN

Specifications

Melting Point 55°C to 58°C
Boiling Point 136°C to 139°C (30 mmHg)
Flash Point 133°C (271°F)
Quantity 5 g
Beilstein 1732524
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material 6-Amino-1-hexanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-748-1
RTECSNumber MO8840000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only