missing translation for 'onlineSavingsMsg'
Learn More

6-amino-1,3-benzoxazol-2(3h)-one, 95%, Thermo Scientific™

Catalog No. CC75714CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC75714CB 250mg Amber Glass Bottle
CC75714DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC75714CB Supplier Thermo Scientific Chemicals Supplier No. CC75714CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22876-17-1
Molecular Formula C7H6N2O2
Molecular Weight (g/mol) 150.137
MDL Number MFCD00463899
InChI Key STLPJYGZOIEDAJ-UHFFFAOYSA-N
Synonym 6-aminobenzo d oxazol-2 3h-one, 6-amino-1,3-benzoxazol-2 3h-one, 2 3h-benzoxazolone, 6-amino, 6-amino-2-3h benzoxazolone, 6-amino-3h-benzooxazol-2-one, 6-amino-2,3-dihydro-1,3-benzoxazol-2-one, 6-amino-2-benzoxazolone, 6-amino-2-benzoxazolinone, 6-amino-3-hydrobenzoxazol-2-one, 6-amino-2-hydroxybenzoxazole
PubChem CID 826989
IUPAC Name 6-amino-3H-1,3-benzoxazol-2-one
SMILES C1=CC2=C(C=C1N)OC(=O)N2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 150.14
Percent Purity 95%
Chemical Name or Material 6-Amino-1, 3-benzoxazol-2(3H)-one
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.