missing translation for 'onlineSavingsMsg'
Learn More

6-Acetoxy-7-methoxy-3H-quinazolin-4-one 98.0+%, TCI America™

Catalog No. p-7118037
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27171G 1 g
A27175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27171G Supplier TCI America Supplier No. A27171G
Only null left

Chemical Identifiers

CAS 179688-53-0
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.211
MDL Number MFCD09951978
InChI Key SOLQIFINSOHAQD-UHFFFAOYSA-N
Synonym 6-Acetoxy-7-methoxy-4(3H)-quinazolinone, 6-Acetoxy-7-methoxy-3,4-dihydroquinazolin-4-one, 6-Acetoxy-7-methoxy-4-hydroxyquinazoline, 6-Acetoxy-7-methoxy-4-quinazolinol
PubChem CID 22022164
IUPAC Name (7-methoxy-4-oxo-1H-quinazolin-6-yl) acetate
SMILES CC(=O)OC1=C(C=C2C(=C1)C(=O)N=CN2)OC

Specifications

Melting Point 297°C
Color Yellow
Quantity 1 g
Formula Weight 234.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Acetoxy-7-methoxy-3H-quinazolin-4-one