missing translation for 'onlineSavingsMsg'
Learn More

6,9-Dichloro-2-methoxyacridine, 98%

Catalog No. AAL0591306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0591306 5 g
AAL0591314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0591306 Supplier Thermo Scientific Chemicals Supplier No. L0591306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-38-4 | C14H9Cl2NO | 278.132 g/mol

6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N'-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N'-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N′-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N′-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 86-38-4
Molecular Formula C14H9Cl2NO
Molecular Weight (g/mol) 278.132
MDL Number MFCD00005028
InChI Key RYRNQWYNHLLOGX-UHFFFAOYSA-N
Synonym halocrin, 3,9-dichloro-7-methoxyacridine, halocrine, 6.9-dichloro-2-methoxyacridine, 2-methoxy-6,9-dichloroacridine, acridine,9-dichloro-2-methoxy, acridine, 6,9-dichloro-2-methoxy, bidd:gt0206, 6,9dichloro-2-methoxyacridine, 6,9-dichloro-2-methoxy-acridine
PubChem CID 66577
IUPAC Name 6,9-dichloro-2-methoxyacridine
SMILES COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl

Specifications

Melting Point 163°C to 165°C
Quantity 5 g
Beilstein 226309
Solubility Information Insoluble in water.
Formula Weight 278.14
Percent Purity 98%
Chemical Name or Material 6,9-Dichloro-2-methoxyacridine
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 201-666-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only