missing translation for 'onlineSavingsMsg'
Learn More

6,8-Dichlorochromone-3-carbonitrile 98.0+%, TCI America™

Catalog No. C31785G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31785G 5 g
C31781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31785G Supplier TCI America Supplier No. C31785G
Only null left

Chemical Identifiers

CAS 72798-32-4
Molecular Formula C10H3Cl2NO2
Molecular Weight (g/mol) 240.039
MDL Number MFCD00192004
InChI Key YERQUFAYRCSXEQ-UHFFFAOYSA-N
Synonym 6,8-Dichloro-3-cyanochromone, 6,8-Dichloro-4-oxo-4H-1-benzopyran-3-carbonitrile
PubChem CID 736020
IUPAC Name 6,8-dichloro-4-oxochromene-3-carbonitrile
SMILES C1=C(C=C2C(=C1Cl)OC=C(C2=O)C#N)Cl

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 5 g
Formula Weight 240.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,8-Dichlorochromone-3-carbonitrile