missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxyquinazoline-2,4-dione 98.0+%, TCI America™

Catalog No. d18395g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D18395G 5 g
D183925G 25 g
2 options

Catalog No. D18395G

Supplier: TCI America D18395G

Only null left

Chemical Identifiers

CAS 28888-44-0
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.20
MDL Number MFCD00023889
InChI Key KWNQIIMVPSMYEM-UHFFFAOYSA-N
Synonym 6,7-dimethoxyquinazoline-2,4-dione, 2,4-dihydroxy-6,7-dimethoxyquinazoline, 6,7-dimethoxyquinazoline-2,4 1h,3h-dione, 6,7-dimethoxyquinazoline-2,4-diol, 6,7-dimethoxy-1,2,3,4-tetrahydroquinazoline-2,4-dione, 6,7-dimethoxy-1h,3h-2,4-quinazolindione, 2,4 1h,3h-quinazolinedione, 6,7-dimethoxy, 6,7-dimethoxy-2,4-quinazolinedione, 2,4-dihydroxy-6,7-dimethoxy quinazoline, 6,7-dimethoxy-2,4-dihydroxy quinazoline
PubChem CID 120081
IUPAC Name 6,7-dimethoxy-1,2,3,4-tetrahydroquinazoline-2,4-dione
SMILES COC1=C(OC)C=C2C(=O)NC(=O)NC2=C1

Specifications

Quantity 5 g
Formula Weight 222.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dimethoxyquinazoline-2,4-dione