missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-4-hydroxyquinoline 98.0+%, TCI America™

Catalog No. D46951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46951G 1 g
D46955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46951G Supplier TCI America Supplier No. D46951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13425-93-9
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD07368026
InChI Key QOGPNCUTXVZQSL-UHFFFAOYSA-N
Synonym 6,7-Dimethoxy-4-quinolinol
PubChem CID 459611
IUPAC Name 6,7-dimethoxy-1H-quinolin-4-one
SMILES COC1=C(C=C2C(=C1)C(=O)C=CN2)OC

Specifications

Melting Point 229°C
Color Yellow
Quantity 1 g
Formula Weight 205.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dimethoxy-4-hydroxyquinoline