missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-3H-quinazolin-4-one 98.0+%, TCI America™

Catalog No. D38165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38165G 5 g
D38161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38165G Supplier TCI America Supplier No. D38165G
Only null left

Chemical Identifiers

CAS 13794-72-4
Molecular Formula C10H10N2O3
Molecular Weight (g/mol) 206.20
MDL Number MFCD01570147
InChI Key DMSRMHGCZUXCMJ-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-3,4-dihydroquinazolin-4-one, 6,7-dimethoxyquinazolin-4 3h-one, 6,7-dimethoxyquinazolin-4-ol, 6,7-dimethoxy-3,4-dihydroquinazoline-4-one, 6,7-dimethoxy-3h-quinazolin-4-one, 6,7-dimethoxy-4 1h-quinazolinone, 6,7-dimethoxy-4 3h-quinazolinone, 6,7-dimethoxyquinazolin-4 1h-one, 6,7-dimethoxy-4-quinazolone, 6,7-dimethoxy-quinazolin-4-ol
PubChem CID 1501878
IUPAC Name 6,7-dimethoxy-1,4-dihydroquinazolin-4-one
SMILES COC1=C(OC)C=C2C(=O)N=CNC2=C1

Specifications

Melting Point 298°C
Color White-Yellow
Quantity 5 g
Formula Weight 206.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dimethoxy-3H-quinazolin-4-one