missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-3,4-dihydroisoquinoline Hydrochloride 98.0+%, TCI America™

Catalog No. p-7128377
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46291G 1 g
D46295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46291G Supplier TCI America Supplier No. D46291G
Only null left

Chemical Identifiers

CAS 20232-39-7
Molecular Formula C11H14ClNO2
Molecular Weight (g/mol) 227.69
MDL Number MFCD11099370
InChI Key PQXVEYYRJHMTEV-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, chloride, 6,7-dimethoxy-3,4-dihydro-isoquinoline hydrochloride, pubchem17358, ksc496s5h, 3,4-dihydro-6,7-dimethoxyisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hydro-chloride, 6,7-dimethoxy-3,4-dihydroisoquinolinehydrochloridehydrate
PubChem CID 2724664
IUPAC Name 6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride
SMILES Cl.COC1=CC2=C(C=NCC2)C=C1OC

Specifications

Melting Point 194°C
Color Yellow
Quantity 1 g
Formula Weight 227.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dimethoxy-3,4-dihydroisoquinoline Hydrochloride
RTECSNumber NW7625000
TSCA No