missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydroxy-4-methylcoumarin, 97%

Catalog No. p-7081765
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1190206 5 g
AAA1190214 25 g
AAA1190222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1190206 Supplier Thermo Scientific Chemicals Supplier No. A1190206
Only null left

CAS: 529-84-0 | C10H8O4 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 529-84-0
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00006859
InChI Key KVOJTUXGYQVLAJ-UHFFFAOYSA-N
Synonym 4-methylesculetin, 6,7-dihydroxy-4-methylcoumarin, methylesculetin, 6,7-dihydroxy-4-methyl-2h-chromen-2-one, 4-methylaesculetin, 4-methylesculetol, 4-methyl-6,7-dihydroxycoumarin, 2h-1-benzopyran-2-one, 6,7-dihydroxy-4-methyl, coumarin, 6,7-dihydroxy-4-methyl, 6,7-dihydroxy-4-methyl-2h-benzopyran-2-one
PubChem CID 5319502
IUPAC Name 6,7-dihydroxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=CC(=C(C=C12)O)O

Specifications

Melting Point ∼275°C (decomposition)
Quantity 5 g
Beilstein 162550
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 6,7-Dihydroxy-4-methylcoumarin
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 208-470-0
RTECSNumber GN6384500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only